siRNA Details
| Gene Symbol: Gm11961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044388.1 | - | 2 | 290 | ||
| CN455772.1 | - | 3 | 318 | |||
| CO427532.1 | - | 1 | 213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11961| Gene Symbol: Gm11961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044388.1 | - | 2 | 290 | ||
| CN455772.1 | - | 3 | 318 | |||
| CO427532.1 | - | 1 | 213 | |||