siRNA Details
| Gene Symbol: Ubtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173784.3 | NP_776145.2 | 1, 2 | 181 | ||
| GenBank mRNA | AK076953.1 | - | 1, 2 | 183 | ||
| BC057026.1 | - | 1, 2 | 181 | 6825513 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubtd2| Gene Symbol: Ubtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173784.3 | NP_776145.2 | 1, 2 | 181 | ||
| GenBank mRNA | AK076953.1 | - | 1, 2 | 183 | ||
| BC057026.1 | - | 1, 2 | 181 | 6825513 | ||