siRNA Details
| Gene Symbol: Zfp616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177570.1 | NP_001171041.1 | 4 | 1499 | ||
| GenBank mRNA | BC083001.1 | - | 4 | 1501 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp616| Gene Symbol: Zfp616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177570.1 | NP_001171041.1 | 4 | 1499 | ||
| GenBank mRNA | BC083001.1 | - | 4 | 1501 | ||