siRNA Details
| Gene Symbol: Gm5082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145878.1 | NP_001139350.1 | 3 | 909 | ||
| NM_177808.4 | NP_808476.2 | 5 | 1072 | |||
| GenBank mRNA | AK037195.1 | - | 5 | 1072 | ||
| BC098182.1 | - | 3 | 909 | |||
| BC147093.1 | - | 1 | 558 | 9056161 | ||
| BC147094.1 | - | 1 | 558 | 9056156 | ||

Gm5082
Genomic Map