siRNA Details
| Gene Symbol: Slc25a48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177809.4 | NP_808477.2 | 6 | 845 | ||
| GenBank mRNA | AK054181.1 | - | 6 | 846 | ||
| BC107250.1 | - | 6 | 809 | 40052351 | ||
| BC107251.1 | - | 6 | 809 | 40052352 | ||
| DV649007.1 | - | 3 | 456 | |||

Slc25a48
Genomic Map