siRNA Details
| Gene Symbol: Gm5084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037190.1 | - | 3 | 2275 | ||
| BC134405.1 | - | 1 | 624 | 40142442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5084| Gene Symbol: Gm5084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037190.1 | - | 3 | 2275 | ||
| BC134405.1 | - | 1 | 624 | 40142442 | ||