siRNA Details
| Gene Symbol: ABCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122674.1 | NP_001116146.1 | 2 | 240 | ||
| NM_002858.3 | NP_002849.1 | 2 | 240 | |||
| XM_006710802.2 | XP_006710865.2 | 3 | 603 | |||
| XM_017001952.1 | XP_016857441.1 | 2 | 781 | |||
| XM_017001953.1 | XP_016857442.1 | 2 | 161 | |||
| GenBank mRNA | AK296791.1 | - | 3 | 276 | ||
| AK308159.1 | - | 2 | 201 | |||
| BC009712.2 | - | 2 | 178 | |||
| BC068509.1 | - | 2 | 201 | 5265201 | ||
| BT006644.1 | - | 2 | 138 | |||
| DC317792.1 | - | 2 | 318 | |||
| M81182.1 | - | 2 | 138 | |||

ABCD3
Genomic Map