siRNA Details
| Gene Symbol: Gm5087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083193.1 | - | 4 | 420 | ||
| BC147066.1 | - | 2 | 249 | 9056061 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5087| Gene Symbol: Gm5087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083193.1 | - | 4 | 420 | ||
| BC147066.1 | - | 2 | 249 | 9056061 | ||