siRNA Details
Gene Symbol: Gm5087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083193.1 | - | 4 | 652 | ||
BC147066.1 | - | 2 | 481 | 9056061 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083193.1 | - | 4 | 652 | ||
BC147066.1 | - | 2 | 481 | 9056061 |