siRNA Details
| Gene Symbol: Rnase9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183032.2 | NP_898853.2 | 2 | 593 | ||
| GenBank mRNA | AY226989.1 | - | 2 | 594 | ||
| AY303816.1 | - | 2 | 593 | |||
| BC107218.1 | - | 2 | 598 | 40051762 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase9| Gene Symbol: Rnase9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183032.2 | NP_898853.2 | 2 | 593 | ||
| GenBank mRNA | AY226989.1 | - | 2 | 594 | ||
| AY303816.1 | - | 2 | 593 | |||
| BC107218.1 | - | 2 | 598 | 40051762 | ||