siRNA Details
| Gene Symbol: Vwa5b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144953.1 | NP_001138425.1 | 7 | 875 | ||
| NM_182636.4 | NP_872574.2 | 7 | 875 | |||
| GenBank mRNA | AK141799.1 | - | 7 | 876 | ||
| BC052055.1 | - | 6 | 821 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa5b2| Gene Symbol: Vwa5b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144953.1 | NP_001138425.1 | 7 | 875 | ||
| NM_182636.4 | NP_872574.2 | 7 | 875 | |||
| GenBank mRNA | AK141799.1 | - | 7 | 876 | ||
| BC052055.1 | - | 6 | 821 | |||