siRNA Details
| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177375.1 | NP_796349.1 | 5 | 616 | ||
| GenBank mRNA | AB232621.2 | - | 5 | 452 | ||
| AK043912.1 | - | 5 | 581 | |||
| BC094931.1 | - | 5 | 524 | 6436242 | ||
| BC118018.1 | - | 5 | 757 | 40103722 | ||

Rab26
Genomic Map