siRNA Details
| Gene Symbol: Treml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033405.2 | NP_001028577.2 | 3 | 1097 | ||
| GenBank mRNA | AK037593.1 | - | 3 | 572 | ||
| DQ087184.1 | - | 3 | 621 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Treml2| Gene Symbol: Treml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033405.2 | NP_001028577.2 | 3 | 1097 | ||
| GenBank mRNA | AK037593.1 | - | 3 | 572 | ||
| DQ087184.1 | - | 3 | 621 | |||