siRNA Details
| Gene Symbol: Acsbg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039114.1 | NP_001034203.1 | 9 | 1220 | ||
| GenBank mRNA | DQ250679.1 | - | 9 | 1220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acsbg2| Gene Symbol: Acsbg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039114.1 | NP_001034203.1 | 9 | 1220 | ||
| GenBank mRNA | DQ250679.1 | - | 9 | 1220 | ||