siRNA Details
| Gene Symbol: Zfp236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177832.3 | NP_808500.2 | 21, 22 | 3934 | ||
| GenBank mRNA | AK156842.1 | - | 1, 2 | 37 | ||
| BC138187.1 | - | 21, 22 | 3934 | 8861206 | ||
| BC145179.1 | - | 22, 23 | 4078 | |||
| BC145180.1 | - | 23, 24 | 4189 | |||

Zfp236
Genomic Map