siRNA Details
| Gene Symbol: Zfp236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177832.3 | NP_808500.2 | 21 | 3720 | ||
| GenBank mRNA | BC138187.1 | - | 21 | 3720 | 8861206 | |
| BC145179.1 | - | 22 | 3864 | |||
| BC145180.1 | - | 23 | 3975 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp236| Gene Symbol: Zfp236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177832.3 | NP_808500.2 | 21 | 3720 | ||
| GenBank mRNA | BC138187.1 | - | 21 | 3720 | 8861206 | |
| BC145179.1 | - | 22 | 3864 | |||
| BC145180.1 | - | 23 | 3975 | |||