siRNA Details
| Gene Symbol: Zfp516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177464.1 | NP_001170935.1 | 2 | 1379 | ||
| NM_183033.2 | NP_898854.1 | 2 | 1401 | |||
| GenBank mRNA | BC053104.1 | - | 1 | 1192 | 6406544 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp516| Gene Symbol: Zfp516 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177464.1 | NP_001170935.1 | 2 | 1379 | ||
| NM_183033.2 | NP_898854.1 | 2 | 1401 | |||
| GenBank mRNA | BC053104.1 | - | 1 | 1192 | 6406544 | |