siRNA Details
| Gene Symbol: Atg2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194348.3 | NP_919329.2 | 40 | 5763 | ||
| GenBank mRNA | AK129132.1 | - | 40 | 5688 | ||
| AK151782.1 | - | 4 | 553 | |||
| BC022657.1 | - | 3 | 306 | |||
| BC023754.1 | - | 36 | 5050 | |||
| BC056482.1 | - | 26 | 3330 | |||
| BC063264.1 | - | 40 | 5704 | 6409527 | ||

Atg2a
Genomic Map