siRNA Details
| Gene Symbol: Unc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175510.3 | NP_780719.2 | 41 | 6352 | ||
| GenBank mRNA | AK173273.1 | - | 7 | 811 | ||
| FJ210934.1 | - | 41 | 6352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc80| Gene Symbol: Unc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175510.3 | NP_780719.2 | 41 | 6352 | ||
| GenBank mRNA | AK173273.1 | - | 7 | 811 | ||
| FJ210934.1 | - | 41 | 6352 | |||