siRNA Details
| Gene Symbol: Rbm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033408.4 | NP_001028580.1 | 6, 7 | 2072 | ||
| GenBank mRNA | AK133355.1 | - | 6, 7 | 2073 | ||
| BC150950.1 | - | 6, 7 | 2034 | 9087861 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm44| Gene Symbol: Rbm44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033408.4 | NP_001028580.1 | 6, 7 | 2072 | ||
| GenBank mRNA | AK133355.1 | - | 6, 7 | 2073 | ||
| BC150950.1 | - | 6, 7 | 2034 | 9087861 | ||