siRNA Details
| Gene Symbol: Lgr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033409.3 | NP_001028581.1 | 6, 7 | 857 | ||
| GenBank mRNA | AK085901.1 | - | 6, 7 | 815 | ||
| AK137380.1 | - | 6, 7 | 858 | |||
| BC141210.1 | - | 6, 7 | 809 | 9055822 | ||

Lgr6
Genomic Map