siRNA Details
| Gene Symbol: Syt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301370.1 | NP_001288299.1 | 6 | 940 | ||
| NM_181546.3 | NP_853524.1 | 6 | 940 | |||
| GenBank mRNA | AB102947.1 | - | 6 | 863 | ||
| AK045901.1 | - | 6 | 941 | |||
| AK051175.1 | - | 3 | 2614 | |||
| BC141262.1 | - | 6 | 882 | 9055874 | ||
| BC145602.1 | - | 6 | 882 | 9054168 | ||

Syt14
Genomic Map