siRNA Details
| Gene Symbol: Snhg7os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030770.1 | - | 3 | 3665 | ||
| AV310154.2 | - | 3 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg7os| Gene Symbol: Snhg7os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030770.1 | - | 3 | 3665 | ||
| AV310154.2 | - | 3 | 418 | |||