siRNA Details
| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181547.3 | NP_853525.3 | 5, 6 | 433 | ||
| GenBank mRNA | AJ564264.1 | - | 5, 6 | 343 | ||
| AK052926.1 | - | 5, 6 | 425 | |||
| AY280963.1 | - | 5, 6 | 386 | |||
| BC069942.1 | - | 5, 6 | 410 | 30443942 | ||

Nostrin
Genomic Map