siRNA Details
| Gene Symbol: Gm14461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177843.3 | NP_808511.1 | 6 | 1388 | ||
| GenBank mRNA | AK044135.1 | - | 6 | 1389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14461| Gene Symbol: Gm14461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177843.3 | NP_808511.1 | 6 | 1388 | ||
| GenBank mRNA | AK044135.1 | - | 6 | 1389 | ||