siRNA Details
| Gene Symbol: Nol4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001986.2 | NP_001001986.2 | 5 | 748 | ||
| NM_001134300.2 | NP_001127772.1 | 8 | 1533 | |||
| GenBank mRNA | AK139459.1 | - | 5 | 748 | ||
| AK147524.1 | - | 8 | 1535 | |||
| AK157863.1 | - | 5 | 729 | |||
| BC075648.1 | - | 7 | 944 | 30620653 | ||
| BC079552.1 | - | 5 | 690 | |||
| BC094234.1 | - | 4 | 599 | |||

Nol4l
Genomic Map