siRNA Details
| Gene Symbol: Fat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183221.3 | NP_899044.3 | 1 | 870 | ||
| GenBank mRNA | DQ286572.1 | - | 1 | 870 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat4| Gene Symbol: Fat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183221.3 | NP_899044.3 | 1 | 870 | ||
| GenBank mRNA | DQ286572.1 | - | 1 | 870 | ||