siRNA Details
| Gene Symbol: PYGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164716.1 | NP_001158188.1 | 17, 18 | 2929 | ||
| GenBank mRNA | AF066859.1 | - | 19, 20 | 2376 | ||
| AK056607.1 | - | 17, 18 | 2188 | |||
| AK057547.1 | - | 2, 3 | 1728 | |||
| AK299376.1 | - | 15, 16 | 1716 | |||
| AK299461.1 | - | 19, 20 | 2395 | |||
| AK314474.1 | - | 19, 20 | 2380 | |||
| BC126392.1 | - | 19, 20 | 2775 | 8992108 | ||
| BC130514.1 | - | 19, 20 | 2775 | 40146545 | ||
| X03031.1 | - | 3, 4 | 353 | |||

PYGM
Genomic Map