siRNA Details
| Gene Symbol: Fam19a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183224.3 | NP_899047.1 | 4, 5 | 701 | ||
| GenBank mRNA | AK078681.1 | - | 4, 5 | 702 | ||
| AY325122.1 | - | 3, 4 | 303 | |||
| BC118531.1 | - | 4, 5 | 391 | 40095917 | ||

Fam19a3
Genomic Map