siRNA Details
| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177859.3 | NP_808527.2 | 5 | 1509 | ||
| GenBank mRNA | AK029552.1 | - | 5 | 1387 | ||
| AK135678.1 | - | 4 | 469 | |||
| AK161171.1 | - | 4 | 341 | |||
| AK164420.1 | - | 5 | 1512 | |||

Aknad1
Genomic Map