siRNA Details
| Gene Symbol: Fam102b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163567.1 | NP_001157039.1 | 4 | 641 | ||
| GenBank mRNA | AK046595.1 | - | 4 | 307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam102b| Gene Symbol: Fam102b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163567.1 | NP_001157039.1 | 4 | 641 | ||
| GenBank mRNA | AK046595.1 | - | 4 | 307 | ||