siRNA Details
| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 2 | 275 | ||
| GenBank mRNA | AK017238.1 | - | 2 | 247 | ||
| AK039177.1 | - | 2 | 246 | |||
| AK042835.1 | - | 2 | 252 | |||
| AK045429.1 | - | 2 | 276 | |||
| AK077246.1 | - | 2 | 326 | |||
| AK084054.1 | - | 1056 | ||||
| AK133059.1 | - | 2 | 256 | |||

Tmem67
Genomic Map