siRNA Details
| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 13, 14 | 1431 | ||
| GenBank mRNA | AK045429.1 | - | 13, 14 | 1432 | ||
| AK077246.1 | - | 13, 14 | 1478 | |||
| AK133059.1 | - | 13, 14 | 1408 | |||

Tmem67
Genomic Map