siRNA Details
| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 23 | 2375 | ||
| GenBank mRNA | AK045429.1 | - | 23 | 2376 | ||
| AK133059.1 | - | 24 | 2460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem67| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 23 | 2375 | ||
| GenBank mRNA | AK045429.1 | - | 23 | 2376 | ||
| AK133059.1 | - | 24 | 2460 | |||