siRNA Details
| Gene Symbol: AI464131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085515.2 | NP_001078984.1 | 2 | 246 | ||
| GenBank mRNA | AK173117.1 | - | 2 | 228 | ||
| BC137640.1 | - | 2 | 203 | 8860659 | ||
| BC140956.1 | - | 2 | 203 | 9055568 | ||
| BY246173.1 | - | 2 | 203 | |||
| BY283520.1 | - | 2 | 248 | |||

AI464131
Genomic Map