siRNA Details
Gene Symbol: Fam166b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177377.5 | NP_796351.2 | 4 | 595 | ||
GenBank mRNA | AK029435.1 | - | 4 | 596 | ||
AK053208.1 | - | 4 | 596 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam166b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177377.5 | NP_796351.2 | 4 | 595 | ||
GenBank mRNA | AK029435.1 | - | 4 | 596 | ||
AK053208.1 | - | 4 | 596 |