siRNA Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025590.4 | NP_079866.2 | 16 | 1835 | ||
| GenBank mRNA | AF416923.1 | - | 16 | 1773 | ||
| AK031588.1 | - | 16 | 1752 | |||
| AK038546.1 | - | 16 | 1773 | |||
| AK048670.1 | - | 16 | 1835 | |||
| AK052332.1 | - | 16 | 1851 | |||
| AK122350.1 | - | 15 | 1622 | |||
| AK170189.1 | - | 9 | 858 | |||
| AK171255.1 | - | 9 | 859 | |||
| BC042492.1 | - | 16 | 1770 | 4240966 | ||

Acot11
Genomic Map