siRNA Details
| Gene Symbol: Foxo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194060.1 | NP_918949.1 | 1 | 361 | ||
| GenBank mRNA | AJ571690.1 | - | 1 | 361 | ||
| BC086628.1 | - | 1 | 449 | 6810803 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxo6| Gene Symbol: Foxo6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194060.1 | NP_918949.1 | 1 | 361 | ||
| GenBank mRNA | AJ571690.1 | - | 1 | 361 | ||
| BC086628.1 | - | 1 | 449 | 6810803 | ||