siRNA Details
| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130030.1 | NP_001123502.1 | 8, 9 | 1114 | ||
| NM_177866.4 | NP_808534.1 | 10, 11 | 1333 | |||
| GenBank mRNA | AK077145.1 | - | 10, 11 | 1320 | ||
| BC108974.2 | - | 10, 11 | 1313 | 40047424 | ||
| BC108975.1 | - | 10, 11 | 1313 | 40047425 | ||
| EU781510.1 | - | 9, 10 | 1210 | |||
| EU781511.1 | - | 8, 9 | 1089 | |||

Catsper4
Genomic Map