siRNA Details
| Gene Symbol: Fhad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177868.4 | NP_808536.2 | 19 | 2588 | ||
| GenBank mRNA | AK148490.1 | - | 4 | 673 | ||
| BC038692.1 | - | 1 | 171 | 1496088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fhad1| Gene Symbol: Fhad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177868.4 | NP_808536.2 | 19 | 2588 | ||
| GenBank mRNA | AK148490.1 | - | 4 | 673 | ||
| BC038692.1 | - | 1 | 171 | 1496088 | ||