siRNA Details
| Gene Symbol: Pramef25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126315.2 | NP_001119787.1 | 3 | 1237 | ||
| Gene Symbol: Gm13119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034101.2 | NP_001029273.2 | 4 | 1233 | ||
| GenBank mRNA | BC100406.1 | - | 3 | 1233 | 30727972 | |

Pramef25
Genomic Map