siRNA Details
| Gene Symbol: Fam185a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177869.4 | NP_808537.2 | 4, 5 | 1024 | ||
| GenBank mRNA | AK030922.1 | - | 4, 5 | 969 | ||
| BC055342.1 | - | 4, 5 | 863 | 6810783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam185a| Gene Symbol: Fam185a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177869.4 | NP_808537.2 | 4, 5 | 1024 | ||
| GenBank mRNA | AK030922.1 | - | 4, 5 | 969 | ||
| BC055342.1 | - | 4, 5 | 863 | 6810783 | ||