siRNA Details
| Gene Symbol: Slc5a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177621.1 | NP_001171092.1 | 5 | 1164 | ||
| NM_001177622.1 | NP_001171093.1 | 5 | 946 | |||
| NM_177870.5 | NP_808538.1 | 4 | 1053 | |||
| GenBank mRNA | AK088682.1 | - | 5 | 852 | ||
| AK158896.1 | - | 5 | 1165 | |||
| AK161633.1 | - | 4 | 977 | |||
| AY572835.1 | - | 2 | 404 | |||
| BC065061.1 | - | 5 | 946 | |||
| BC085132.1 | - | 4 | 749 | 6416422 | ||
| BC117852.1 | - | 4 | 706 | 40094012 | ||
| BC117853.1 | - | 4 | 706 | 40094013 | ||

Slc5a6
Genomic Map