siRNA Details
| Gene Symbol: Miat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB294520.1 | - | 1, 2 | 91 | ||
| AB294521.1 | - | 1, 2 | 91 | |||
| AB294522.1 | - | 1, 2 | 91 | |||
| AB294525.1 | - | 1, 2 | 91 | |||
| AB294526.1 | - | 1, 2 | 91 | |||
| AB294527.1 | - | 1, 2 | 91 | |||
| AB300594.1 | - | 1, 2 | 328 | |||
| AK038726.1 | - | 1, 2 | 342 | |||

Miat
Genomic Map