siRNA Details
| Gene Symbol: 2610524H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181075.3 | NP_851420.1 | 2 | 241 | ||
| GenBank mRNA | AK012147.1 | - | 2 | 230 | ||
| AK148078.1 | - | 2 | 216 | |||
| BC089033.1 | - | 2 | 221 | |||
| BC096574.1 | - | 2 | 232 | 1495259 | ||
| BY090347.1 | - | 2 | 241 | |||

2610524H06Rik
Genomic Map