siRNA Details
| Gene Symbol: Tmem120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039723.2 | NP_001034812.1 | 8 | 799 | ||
| GenBank mRNA | AK132283.1 | - | 8 | 768 | ||
| AK158123.1 | - | 8 | 758 | |||
| AK172110.1 | - | 8 | 739 | |||
| BC022593.1 | - | 8 | 750 | 4038363 | ||
| BC115785.1 | - | 7 | 643 | 40104955 | ||
| BC118014.1 | - | 8 | 760 | 40104954 | ||

Tmem120b
Genomic Map