siRNA Details
| Gene Symbol: Tmem120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039723.2 | NP_001034812.1 | 9 | 848 | ||
| GenBank mRNA | AK132283.1 | - | 9 | 817 | ||
| AK158123.1 | - | 9 | 807 | |||
| BC022593.1 | - | 9 | 799 | 4038363 | ||
| BC115785.1 | - | 8 | 692 | 40104955 | ||
| BC118014.1 | - | 9 | 809 | 40104954 | ||

Tmem120b
Genomic Map