siRNA Details
| Gene Symbol: Sdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177879.5 | NP_808547.3 | 23, 24 | 3696 | ||
| GenBank mRNA | AK147578.1 | - | 23, 24 | 3741 | ||
| AY353236.1 | - | 19, 20 | 2846 | |||
| BC060237.1 | - | 23, 24 | 3280 | |||
| BC138453.1 | - | 19, 20 | 2717 | 8861473 | ||

Sdk1
Genomic Map