siRNA Details
| Gene Symbol: Fam71f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289663.1 | NP_001276592.1 | 3 | 636 | ||
| NM_001289664.1 | NP_001276593.1 | 3 | 636 | |||
| NM_001289665.1 | NP_001276594.1 | 3 | 631 | |||
| NM_207258.3 | NP_997141.2 | 3 | 636 | |||
| GenBank mRNA | AB473725.1 | - | 3 | 577 | ||
| AK133908.1 | - | 3 | 637 | |||
| BC147513.1 | - | 3 | 619 | 9056505 | ||
| BC172074.1 | - | 3 | 619 | 9054360 | ||
| BC172085.1 | - | 3 | 614 | |||
| BC172086.1 | - | 3 | 619 | 9054372 | ||

Fam71f1
Genomic Map