siRNA Details
Gene Symbol: D630045J12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194061.2 | NP_918950.2 | 2 | 913 | ||
GenBank mRNA | BC079889.1 | - | 1 | 275 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D630045J12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194061.2 | NP_918950.2 | 2 | 913 | ||
GenBank mRNA | BC079889.1 | - | 1 | 275 |